Learn More
Abnova™ Human ALG13 Full-length ORF (NP_060936.1, 1 a.a. - 165 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for Array, ELISA, WB-Re
Brand: Abnova™ H00079868-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Asparagine (N)-glycosylation is an essential modification that regulates protein folding and stability. ALG13 and ALG14 (MIM 612866) constitute the UDP-GlcNAc transferase, which catalyzes a key step in endoplasmic reticulum N-linked glycosylation (Averbeck et al., 2007 [PubMed 17686769]).[supplied by OMIM]
Sequence: MKCVFVTVGTTSFDDLIACVSAPDSLQKIESLGYNRLILQIGRGTVVPEPFSTESFTLDVYRYKDSLKEDIQKADLVISHAGAGSCLETLEKGKPLVVVINEKLMNNHQLELAKQLHKEGHLFYCTCSTLPGLLQSMDLSTLKCYPPGQPEKFSAFLDKVVGLQKSpecifications
NP_060936.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ALG13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Protein Array, ELISA, Western Blot | |
79868 | |
ALG13 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CXorf45/FLJ23018/GLT28D1/MDS031/MGC12423/YGL047W | |
ALG13 | |
Recombinant | |
wheat germ expression system |