Learn More
Abnova™ Human ALG8 Partial ORF (NP_076984, 260 a.a. - 334 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079053-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the ALG6/ALG8 glucosyltransferase family. The encoded protein catalyzes the addition of the second glucose residue to the lipid-linked oligosaccharide precursor for N-linked glycosylation of proteins. Mutations in this gene have been associated with congenital disorder of glycosylation type Ih (CDG-Ih). Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: NQLPQVFSRLFPFKRGLCHAYWAPNFWALYNALDKVLSVIGLKLKFLDPNNIPKASMTSGLVQQFQHTVLPSVTPSpecifications
NP_076984 | |
Liquid | |
79053 | |
ALG8 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC2840 | |
ALG8 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.99kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NQLPQVFSRLFPFKRGLCHAYWAPNFWALYNALDKVLSVIGLKLKFLDPNNIPKASMTSGLVQQFQHTVLPSVTP | |
RUO | |
ALG8 | |
Wheat Germ (in vitro) | |
GST |