Learn More
Abnova™ Human ALOX15B Full-length ORF (AAH35217.1, 1 a.a. - 676 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000247-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the lipoxygenase family of structurally related nonheme iron dioxygenases involved in the production of fatty acid hydroperoxides. The encoded protein converts arachidonic acid exclusively to 15S-hydroperoxyeicosatetraenoic acid, while metabolizing linoleic acid less effectively. This gene is located in a cluster of related genes and a pseudogene that spans approximately 100 kilobases on the short arm of chromosome 17. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]
Sequence: MAEFRVRVSTGEAFGAGTWDKVSVSIVGTRGESPPLPLDNLGKEFTAGAEEDFQVTLPEDVGRVLLLRVHKAPPVLPLLGPLAPDAWFCRWFQLTPPRGGHLLFPCYQWLEGAGTLVLQEGTAKVSWADHHPVLQQQRQEELQARQEMYQWKAYNPGWPHCLDEKTVEDLELNIKYSTAKNANFYLQAGSAFAEMKIKGLLDRKGLWRSLNEMKRIFNFRRTPAAEHAFEHWQEDAFFASQFLNGLNPVLIRRCHYLPKNFPVTDAMVASVLGPGTSLQAELEKGSLFLVDHGILSGIQTNVINGKPQFSAAPMTLLYQSPGCGPLLPLAIQLSQTPGPNSPIFLPTDDKWDWLLAKTWVRNAEFSFHEALTHLLHSHLLPEVFTLATLRQLPHCHPLFKLLIPHTRYTLHINTLARELLIVPGQVVDRSTGIGIEGFSELIQRNMKQLNYSLLCLPEDIRTRGVEDIPGYYYRDDGMQIWGAVERFVSEIIGIYYPSDESVQDDRELQAWVREIFSKGFLNQESSGIPSSLETREALVQYVTMVIFTCSAKHAAVSAGQFDSCAWMPNLPPSMQLPPPTSKGLATCEGFIATLPPVNATCDVILALWLLSKEPGDQRPLGTYPDEHFTEEAPRRSIATFQSRLAQISRGIQERNRGLVLPYTYLDPPLIENSVSISpecifications
AAH35217.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
102.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ALOX15B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
247 | |
ALOX15B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
15-LOX-2 | |
ALOX15B | |
Recombinant | |
wheat germ expression system |