Learn More
Abnova™ Human ALOX15B Full-length ORF (AAH35217.1, 1 a.a. - 676 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH35217.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 247 |
Molecular Weight (g/mol) | 102.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16181511
|
Abnova™
H00000247-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16171511
|
Abnova™
H00000247-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the lipoxygenase family of structurally related nonheme iron dioxygenases involved in the production of fatty acid hydroperoxides. The encoded protein converts arachidonic acid exclusively to 15S-hydroperoxyeicosatetraenoic acid, while metabolizing linoleic acid less effectively. This gene is located in a cluster of related genes and a pseudogene that spans approximately 100 kilobases on the short arm of chromosome 17. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]
Sequence: MAEFRVRVSTGEAFGAGTWDKVSVSIVGTRGESPPLPLDNLGKEFTAGAEEDFQVTLPEDVGRVLLLRVHKAPPVLPLLGPLAPDAWFCRWFQLTPPRGGHLLFPCYQWLEGAGTLVLQEGTAKVSWADHHPVLQQQRQEELQARQEMYQWKAYNPGWPHCLDEKTVEDLELNIKYSTAKNANFYLQAGSAFAEMKIKGLLDRKGLWRSLNEMKRIFNFRRTPAAEHAFEHWQEDAFFASQFLNGLNPVLIRRCHYLPKNFPVTDAMVASVLGPGTSLQAELEKGSLFLVDHGILSGIQTNVINGKPQFSAAPMTLLYQSPGCGPLLPLAIQLSQTPGPNSPIFLPTDDKWDWLLAKTWVRNAEFSFHEALTHLLHSHLLPEVFTLATLRQLPHCHPLFKLLIPHTRYTLHINTLARELLIVPGQVVDRSTGIGIEGFSELIQRNMKQLNYSLLCLPEDIRTRGVEDIPGYYYRDDGMQIWGAVERFVSEIIGIYYPSDESVQDDRELQAWVREIFSKGFLNQESSGIPSSLETREALVQYVTMVIFTCSAKHAAVSAGQFDSCAWMPNLPPSMQLPPPTSKGLATCEGFIATLPPVNATCDVILALWLLSKEPGDQRPLGTYPDEHFTEEAPRRSIATFQSRLAQISRGIQERNRGLVLPYTYLDPPLIENSVSISpecifications
AAH35217.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
102.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
15-LOX-2 | |
ALOX15B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
247 | |
ALOX15B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ALOX15B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |