missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human AMOTL2 Full-length ORF (AAH11454.1, 1 a.a. - 466 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH11454.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51421 |
Molecular Weight (g/mol) | 78.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16180003
|
Abnova™
H00051421-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16190003
|
Abnova™
H00051421-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Angiomotin is a protein that binds angiostatin, a circulating inhibitor of the formation of new blood vessels (angiogenesis). Angiomotin mediates angiostatin inhibition of endothelial cell migration and tube formation in vitro. The protein encoded by this gene is related to angiomotin and is a member of the motins protein family. [provided by RefSeq]
Sequence: MEAVLRENARLQRDNERLQRELESSAEKAGRIEKLESEIQRLSEAHESLTRASSKREALEKTMRNKMDSEMRRLQDFNRDLRERLESANRRLASKTQEAQAGSQDMVAKLLAQSYEQQQEQEKLEREMALLRGAIEDQRRRAELLEQALGNAQGRAARAEEELRKKQAYVEKVERLQQALGQLQAACEKREQLELRLRTRLEQELKALRAQQRQAGAPGGSSGSGGSPELSALRLSEQLREKEEQILALEADMTKWEQKYLEERAMRQFAMDAAATAAAQRDTTLIRHSPQPSPSSSFNEGLLTGGHRHQEMESRLKVLHAQILEKDAVIKVLQQRSRRDPGKAIQGSLRPAKSVPSVFAAAAAGTQGWQGLSSSERQTADAPARLTTDRAPTEEPVVTAPPAAHAKHGSRDGSTQTEGPPDSTSTCLPPEPDSLLGCSSSQRAASLDSVATSRVQDLSDMVEILISpecifications
AAH11454.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
78.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LCCP | |
AMOTL2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
51421 | |
AMOTL2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
AMOTL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |