Learn More
Abnova™ Human AMOTL2 Full-length ORF (AAH11454.1, 1 a.a. - 466 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051421-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Angiomotin is a protein that binds angiostatin, a circulating inhibitor of the formation of new blood vessels (angiogenesis). Angiomotin mediates angiostatin inhibition of endothelial cell migration and tube formation in vitro. The protein encoded by this gene is related to angiomotin and is a member of the motins protein family. [provided by RefSeq]
Sequence: MEAVLRENARLQRDNERLQRELESSAEKAGRIEKLESEIQRLSEAHESLTRASSKREALEKTMRNKMDSEMRRLQDFNRDLRERLESANRRLASKTQEAQAGSQDMVAKLLAQSYEQQQEQEKLEREMALLRGAIEDQRRRAELLEQALGNAQGRAARAEEELRKKQAYVEKVERLQQALGQLQAACEKREQLELRLRTRLEQELKALRAQQRQAGAPGGSSGSGGSPELSALRLSEQLREKEEQILALEADMTKWEQKYLEERAMRQFAMDAAATAAAQRDTTLIRHSPQPSPSSSFNEGLLTGGHRHQEMESRLKVLHAQILEKDAVIKVLQQRSRRDPGKAIQGSLRPAKSVPSVFAAAAAGTQGWQGLSSSERQTADAPARLTTDRAPTEEPVVTAPPAAHAKHGSRDGSTQTEGPPDSTSTCLPPEPDSLLGCSSSQRAASLDSVATSRVQDLSDMVEILISpecifications
AAH11454.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
78.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
AMOTL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51421 | |
AMOTL2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LCCP | |
AMOTL2 | |
Recombinant | |
wheat germ expression system |