missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human APIP Full-length ORF (AAH17594.1, 1 a.a. - 242 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051074-P01.10ug
Additional Details : Weight : 0.00010kg
Description
APIP is an APAF1 (MIM 602233)-interacting protein that acts as a negative regulator of ischemic/hypoxic injury (Cho et al., 2004 [PubMed 15262985]).[supplied by OMIM]
Sequence: MSGCDAWEGDCCSRRCGAQDKEHPRYLIPELCKQFYHLGWVTGTGGGISLKHGDEIYIAPSGVQKERIQPEDMFVYDINEKDISGPSPSKKLKKSQCTPLFMNAYTMRGAGAVIHTHSKAAVMATLLFPGREFKITHQEMIKGIKKCTSGGYYRYDDMLVVPIIENTPEEKDLKDRMAHAMNEYPDSCAVLVRRHGVYVWGETWEKAKTMCECYDYLFDIAVSMKKVGLDPSQLPVGENGIVSpecifications
AAH17594.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
53.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
APIP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51074 | |
APIP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APIP2/CGI-29/CGI29/MMRP19/dJ179L10.2 | |
APIP | |
Recombinant | |
wheat germ expression system |