missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human APOBEC1 Partial ORF (NP_001635, 137 a.a. - 236 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000339-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16184754

  • 3343.96 SEK / 10µg

Estimated Shipment: 15-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the cytidine deaminase enzyme family. The encoded protein forms a multiple-protein editing holoenzyme with APOBEC1 complementation factor (ACF) and APOBEC1 stimulating protein (ASP). This holoenzyme is involved in the editing of C-to-U nucleotide bases in apolipoprotein B and neurofibromatosis-1 mRNAs. [provided by RefSeq]

Sequence: SGVTIQIMRASEYYHCWRNFVNYPPGDEAHWPQYPPLWMMLYALELHCIILSLPPCLKISRRWQNHLTFFRLHLQNCHYQTIPPHILLATGLIHPSVAWR
Specifications

Specifications

NP_001635
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SGVTIQIMRASEYYHCWRNFVNYPPGDEAHWPQYPPLWMMLYALELHCIILSLPPCLKISRRWQNHLTFFRLHLQNCHYQTIPPHILLATGLIHPSVAWR
RUO
APOBEC1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
339
APOBEC1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
APOBEC-1/BEDP/CDAR1/HEPR
APOBEC1
Yes
Videos
SDS
Documents
Certificates