missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human APOBEC3F Full-length ORF (NP_001006667.1, 1 a.a. - 101 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00200316-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16149233

  • 6845.00 SEK / 25µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the cytidine deaminase gene family. It is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MKPHFRNTVERMYRDTFSYNFYNRPILSRRNTVWLCYEVKTKGPSRPRLDAKIFRGQVPRSFIRAPFQVLSSPFGQCAPPHGTAQVQWPPQLTAGREQGRP
Specifications

Specifications

NP_001006667.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
APOBEC3F
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
200316
APOBEC3F (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARP8/BK150C2.4.MRNA/KA6/MGC74891
APOBEC3F
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers