Learn More
Abnova™ Human APOBEC3F Full-length ORF (NP_001006667.1, 1 a.a. - 101 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00200316-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the cytidine deaminase gene family. It is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MKPHFRNTVERMYRDTFSYNFYNRPILSRRNTVWLCYEVKTKGPSRPRLDAKIFRGQVPRSFIRAPFQVLSSPFGQCAPPHGTAQVQWPPQLTAGREQGRPSpecifications
NP_001006667.1 | |
Liquid | |
200316 | |
APOBEC3F (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARP8/BK150C2.4.MRNA/KA6/MGC74891 | |
APOBEC3F | |
Yes | |
wheat germ expression system |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
38.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
APOBEC3F | |
Wheat Germ (in vitro) | |
GST |