missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human Apolipoprotein B/ApoB Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB41242-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Apolipoprotein B/ApoB Monoclonal specifically detects Apolipoprotein B/ApoB in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifikationer
| Apolipoprotein B/ApoB | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Apo B-100, APOB, apoB-100, ApoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100 | |
| E. coli-derived recombinant human Apolipoprotein B/ApoB, Ile3374-Val3600, Accession # P04114 | |
| 25 μg | |
| 338 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 1008037 | |
| Western Blot 1 μg/mL, Immunohistochemistry 5-25 μg/mL | |
| P04114 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Apolipoprotein B/ApoB in direct ELISAs. | |
| Human | |
| Lyophilized |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering