Learn More
Abcam Human APPL1 (APPL) knockout HeLa cell line
Product Code. 32141663 Shop All Abcam Products
Change view
Click to view available options
Quantity:
2 x 1,000,000 Cells
Unit Size:
Pack of 2
Product Code. Quantity Unit Size
32141663 2 x 1,000,000 Cells Pack of 2
1 options
Product Code. 32141663 Supplier Abcam Supplier No. ab2651872x1000000Cells_
 Restricted Item
Beräknad leverans: 07-12-2026
APPL1 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 2 bp deletion in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockout cells: select '...
APPL1 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 2 bp deletion in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells
Regulatory Status RUO