missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human APRIN Partial ORF (NP_055847, 1168 a.a. - 1259 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023047-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: GRIKGRLDSSEMDHSENEDYTMSSPLPGKKSDKRDDSDLVRSELEKPRGRKKTPVTEQEEKLGMDDLTKLVQEQKPKGSQRSRKRGHTASESSpecifications
NP_055847 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.86kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GRIKGRLDSSEMDHSENEDYTMSSPLPGKKSDKRDDSDLVRSELEKPRGRKKTPVTEQEEKLGMDDLTKLVQEQKPKGSQRSRKRGHTASES | |
RUO | |
PDS5B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23047 | |
APRIN (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APRIN/AS3/CG008/FLJ23236/KIAA0979/RP1-267P19.1 | |
PDS5B | |
Recombinant | |
wheat germ expression system |