missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human APRT Full-length ORF (NP_000476.1, 1 a.a. - 180 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000353-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16171731

  • 6845.00 SEK / 25µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Adenine phosphoribosyltransferase belongs to the purine/pyrimidine phosphoribosyltransferase family. A conserved feature of this gene is the distribution of CpG dinucleotides. This enzyme catalyzes the formation of AMP and inorganic pyrophosphate from adenine and 5-phosphoribosyl-1-pyrophosphate (PRPP). It also produces adenine as a by-product of the polyamine biosynthesis pathway. A homozygous deficiency in this enzyme causes 2,8-dihydroxyadenine urolithiasis. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MADSELQLVEQRIRSFPDFPTPGVVFRDISPVLKDPASFRAAIGLLARHLKATHGGRIDYIAGLDSRGFLFGPSLAQELGLGCVLIRKRGKLPGPTLWASYSLEYGKAELEIQKDALEPGQRVVVVDDLLATGGTMNAACELLGRLQAEVLECVSLVELTSLKGREKLAPVPFFSLLQYE
Specifications

Specifications

NP_000476.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
46kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
APRT
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
353
APRT (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AMP/DKFZp686D13177/MGC125856/MGC125857/MGC129961
APRT
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers