missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARHGEF1 Partial ORF (AAH34013.2, 830 a.a. - 927 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH34013.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9138 |
Molecular Weight (g/mol) | 36.41kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16180026
|
Abnova™
H00009138-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16170026
|
Abnova™
H00009138-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate Rho-dependent signals. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been defined. [provided by RefSeq]
Sequence: CRPGPEGQLAATALRKVLSLKQLLFPAEEDNGAGPPRDGDGVPGGGPLSPARTQEIQENLLSLEETMKQLEELEEEFCRLRPLLSQLGGNSVPQPGCTSpecifications
AAH34013.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GEF1/LBCL2/LSC/P115-RHOGEF/SUB1.5 | |
ARHGEF1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9138 | |
ARHGEF1 (Human) Recombinant Protein (Q01) | |
CRPGPEGQLAATALRKVLSLKQLLFPAEEDNGAGPPRDGDGVPGGGPLSPARTQEIQENLLSLEETMKQLEELEEEFCRLRPLLSQLGGNSVPQPGCT | |
RUO | |
ARHGEF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |