Learn More
Abnova™ Human ARHGEF10 Full-length ORF (AAH40474.1, 1 a.a. - 380 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009639-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate Rho-dependent signals. [provided by RefSeq]
Sequence: MENPEEAIYDDVPRENSDSEPDEMIYDDVENGDEGGNSSLEYGWSSSEFESYEEQSDSECKNGIPRSFLRSNHKKQMQKLVKAAKDGTKDGLERTRAAVKRGRSFIRTKSLIAQDHRSSLEEEQNLFIDVDCKHPEAILTPMPEGLSQQQVVRRYILGSVVDSEKNYVDALKRILEQYEKPLSEMEPKVLSERKLKTVFYRVKEILQCHSLFQIALASRVSEWDSVEMIGVVFVASFSKSMVLDAYSEYVNNFSTAVAVLKKTCATKPAFLEFLKQEQEASPDRTTLYSLMMKPIQRFPQFILLLQDMLKNTSKGHPDRLPLQMALTELETLAEKLNERKRDADQRCEVKQIAKAINERYLNKVERGFLQLYSKIIFALCSpecifications
AAH40474.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ARHGEF10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9639 | |
ARHGEF10 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686H0726/GEF10/MGC131664 | |
ARHGEF10 | |
Recombinant | |
wheat germ expression system |