Learn More
Abnova™ Human ARHGEF18 Full-length ORF (AAH08016.1, 1 a.a. - 89 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023370-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Rho GTPases are GTP binding proteins that regulate a wide spectrum of cellular functions. These cellular processes include cytoskeletal rearrangements, gene transcription, cell growth and motility. Activation of Rho GTPases is under the direct control of guanine nucleotide exchange factors (GEFs). The protein encoded by this gene is a guanine nucleotide exchange factor and belongs to the Rho GTPase GFE family. Family members share a common feature, a Dbl (DH) homology domain followed by a pleckstrin (PH) homology domain. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MSQGMQRMHLETLQQVDKWPLCGPLACSELLQLTVRSLEGWRKEVLGSIKGAGTSQGGEIHPRSSSGGERAHVKPCSAPQALVVGLGPGSpecifications
AAH08016.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ARHGEF18 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
23370 | |
ARHGEF18 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0521/MGC15913/P114-RhoGEF | |
ARHGEF18 | |
Recombinant | |
wheat germ expression system |