Learn More
Abnova™ Human ARHGEF7 Partial ORF (NP_663788, 102 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008874-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Rho GTPases play a fundamental role in numerous cellular processes triggered by extracellular stimuli that work through G protein coupled receptors. The encoded protein belongs to a family of cytoplasmic proteins that activate the Ras-like family of Rho proteins by exchanging bound GDP for GTP. It forms a complex with the small GTP binding protein Rac1 and recruits Rac1 to membrane ruffles and to focal adhesions. This protein can induce membrane ruffling. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]
Sequence: SSLVTLNKVTADIGLGSDSVCARPSSHRIKSFDSLGSQSLHTRTSKLFQGQYRSLDMTDNSNNQLVVRAKFNFQQTNEDELSFSKGDVISpecifications
NP_663788 | |
Liquid | |
8874 | |
ARHGEF7 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BETA-PIX/COOL1/DKFZp686C12170/DKFZp761K1021/KIAA0142/KIAA0412/Nbla10314/P50/P50BP/P85/P85COOL1/P85SPR/PAK3/PIXB | |
ARHGEF7 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SSLVTLNKVTADIGLGSDSVCARPSSHRIKSFDSLGSQSLHTRTSKLFQGQYRSLDMTDNSNNQLVVRAKFNFQQTNEDELSFSKGDVI | |
RUO | |
ARHGEF7 | |
Wheat Germ (in vitro) | |
GST |