missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARSF Full-length ORF (NP_004033.2, 1 a.a. - 590 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_004033.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 416 |
Molecular Weight (g/mol) | 92.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16141921
|
Abnova™
H00000416-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16131921
|
Abnova™
H00000416-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a member of the sulfatase family, and more specifically, the arylsulfatase subfamily. Members of the subfamily share similarity in sequence and splice sites, and are clustered together on chromosome X, suggesting that they are derived from recent gene duplication events. Sulfatases are essential for the correct composition of bone and cartilage matrix. The activity of this protein, unlike that of arylsulfatase E, is not inhibited by warfarin. [provided by RefSeq]
Sequence: MRPRRPLVFMSLVCALLNTCQAHRVHDDKPNIVLIMVDDLGIGDLGCYGNDTMRTPHIDRLAREGVRLTQHISAASLCSPSRSAFLTGRYPIRSGMVSSGNRRVIQNLAVPAGLPLNETTLAALLKKQGYSTGLIGKWHQGLNCDSRSDQCHHPYNYGFDYYYGMPFTLVDSCWPDPSRNTELAFESQLWLCVQLVAIAILTLTFGKLSGWVSVPWLLIFSMILFIFLLGYAWFSSHTSPLYWDCLLMRGHEITEQPMKAERAGSIMVKEAISFLERHSKETFLLFFSFLHVHTPLPTTDDFTGTSKHGLYGDNVEEMDSMVGKILDAIDDFGLRNNTLVYFTSDHGGHLEARRGHAQLGGWNGIYKGGKGMGGWEGGIRVPGIVRWPGKVPAGRLIKEPTSLMDILPTVASVSGGSLPQDRVIDGRDLMPLLQGNVRHSEHEFLFHYCGSYLHAVRWIPKDDSGSVWKAHYVTPVFQPPASGGCYVTSLCRCFGEQVTYHNPPLLFDLSRDPSESTPLTPATEPLHDFVIKKVANALKEHQETIVPVTYQLSELNQGRTWLKPCCGVFPFCLCDKEEEVSQPRGPNEKRSpecifications
NP_004033.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
92.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ASF | |
ARSF | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
416 | |
ARSF (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ARSF | |
Wheat Germ (in vitro) | |
GST | |
Liquid |