missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ASB10 Partial ORF (NP_001135931, 48 a.a. - 153 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_001135931 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 136371 |
Molecular Weight (g/mol) | 37.4kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16121877
|
Abnova™
H00136371-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16111877
|
Abnova™
H00136371-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. The SOCS box serves to couple suppressor of cytokine signaling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene. [provided by RefSeq]
Sequence: MSWSPEECKGQGEPLDDRHPLCARLVEKPSRGSEEHLKSGPGPIVTRTASGPALAFWQAVLAGDVGCVSRILADSSTGLAPDSVFDTSDPERWRDFRFNIRALRLWSpecifications
NP_001135931 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.4kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ASB10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
136371 | |
ASB10 (Human) Recombinant Protein (Q01) | |
MSWSPEECKGQGEPLDDRHPLCARLVEKPSRGSEEHLKSGPGPIVTRTASGPALAFWQAVLAGDVGCVSRILADSSTGLAPDSVFDTSDPERWRDFRFNIRALRLW | |
RUO | |
ASB10 | |
Recombinant | |
wheat germ expression system |