missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ASB5 Partial ORF (NP_543150, 220 a.a. - 328 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_543150 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 140458 |
Molecular Weight (g/mol) | 37.73kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16171907
|
Abnova™
H00140458-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16161907
|
Abnova™
H00140458-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene but their full length sequences are not known. [provided by RefSeq]
Sequence: LLYAGADVQKGKYWDTPLHAAAQQSSTEIVNLLLEFGADINAKNTELLRPIDVATSSSMVERILLQHEATPSSLYQLCRLCIRSYIGKPRLHLIPQLQLPTLLKNFLQYSpecifications
NP_543150 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ASB5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
140458 | |
ASB5 (Human) Recombinant Protein (Q01) | |
LLYAGADVQKGKYWDTPLHAAAQQSSTEIVNLLLEFGADINAKNTELLRPIDVATSSSMVERILLQHEATPSSLYQLCRLCIRSYIGKPRLHLIPQLQLPTLLKNFLQY | |
RUO | |
ASB5 | |
Recombinant | |
wheat germ expression system |