missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ATN1 Partial ORF (AAH51795, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH51795 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1822 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16144191
|
Abnova™
H00001822-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16134191
|
Abnova™
H00001822-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Dentatorubral pallidoluysian atrophy is a rare neurodegenerative disorder characterized by cerebellar ataxia, myoclonic epilepsy, choreoathetosis, and dementia. The disorder is related to the expansion of a trinucleotide repeat within this gene. The encoded protein includes a serine repeat and a region of alternating acidic and basic amino acids, as well as the variable glutamine repeat. Alternative splicing results in two transcripts variants that encode the same protein. [provided by RefSeq]
Sequence: MKTRQNKDSMSMRSGRKKEAPGPREELRSRGRASPGGVSTSSSDGKAEKSRQTAKKARVEEASTPKVNKQGRSEEISESESEETNAPKKTKTEQELPRPQSPSDLDSLDGSpecifications
AAH51795 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
B37/D12S755E/DRPLA/NOD | |
ATN1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1822 | |
ATN1 (Human) Recombinant Protein (Q01) | |
MKTRQNKDSMSMRSGRKKEAPGPREELRSRGRASPGGVSTSSSDGKAEKSRQTAKKARVEEASTPKVNKQGRSEEISESESEETNAPKKTKTEQELPRPQSPSDLDSLDG | |
RUO | |
ATN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |