Learn More
Abnova™ Human ATP7A Partial ORF (NP_000043, 1406 a.a. - 1500 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000538-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a transmembrane protein that functions in copper transport across membranes. The protein localizes to the trans-Golgi network, where it is predicted to supply copper to copper-dependent enzymes in the secretory pathway. The protein relocalizes to the plasma membrane under conditions of elevated extracellular copper and functions in the efflux of copper from cells. Mutations in this gene result in Menkes disease, X-linked cutis laxa, and occipital horn syndrome. [provided by RefSeq]
Sequence: FLKLYRKPTYESYELPARSQIGQKSPSEISVHVGIDDTSRNSPKLGLLDRIVNYSRASINSLLSDKRSLNSVVTSEPDKHSLLVGDFREDDDTALSpecifications
NP_000043 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.19kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FLKLYRKPTYESYELPARSQIGQKSPSEISVHVGIDDTSRNSPKLGLLDRIVNYSRASINSLLSDKRSLNSVVTSEPDKHSLLVGDFREDDDTAL | |
RUO | |
ATP7A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
538 | |
ATP7A (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ17790/MK/MNK | |
ATP7A | |
Recombinant | |
wheat germ expression system |