missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ATP8B4 Partial ORF (NP_079113.2, 401 a.a. - 488 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079895-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16130117

  • 6845.00 SEK / 25µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: IMTFKRCSINGRIYGEVHDDLDQKTEITQEKEPVDFSVKSQADREFQFFDHHLMESIKMGDPKVHEFLRLLALCHTVMSEENSAGELI
Specifications

Specifications

NP_079113.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.42kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
IMTFKRCSINGRIYGEVHDDLDQKTEITQEKEPVDFSVKSQADREFQFFDHHLMESIKMGDPKVHEFLRLLALCHTVMSEENSAGELI
RUO
ATP8B4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
79895
ATP8B4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATPIM
ATP8B4
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers