missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ATPAF2 Partial ORF (NP_663729, 180 a.a. - 289 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_663729 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 91647 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16181167
|
Abnova™
H00091647-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16171167
|
Abnova™
H00091647-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes an assembly factor for the F(1) component of the mitochondrial ATP synthase. This protein binds specifically to the F1 alpha subunit and is thought to prevent this subunit from forming nonproductive homooligomers during enzyme assembly. This gene is located within the Smith-Magenis syndrome region on chromosome 17. An alternatively spliced transcript variant has been described, but its biological validity has not been determined. [provided by RefSeq]
Sequence: MGPSIPAKTREVLVSHLASYNTWALQGIEFVAAQLKSMVLTLGLIDLRLTVEQAVLLSRLEEEYQIQKWGNIEWAHDYELQELRARTAAGTLFIHLCSESTTVKHKLLKESpecifications
NP_663729 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATP12/ATP12p/LP3663/MGC29736 | |
ATPAF2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
91647 | |
ATPAF2 (Human) Recombinant Protein (Q01) | |
MGPSIPAKTREVLVSHLASYNTWALQGIEFVAAQLKSMVLTLGLIDLRLTVEQAVLLSRLEEEYQIQKWGNIEWAHDYELQELRARTAAGTLFIHLCSESTTVKHKLLKE | |
RUO | |
ATPAF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |