missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human AYP1 Full-length ORF (NP_115569.2, 1 a.a. - 164 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_115569.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 84153 |
Molecular Weight (g/mol) | 44.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16195663
|
Abnova™
H00084153-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16185663
|
Abnova™
H00084153-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a ribonuclease H subunit that can cleave ribonucleotides from RNA:DNA duplexes. Mutations in this gene cause Aicardi-Goutieres syndrome-3, a disease that causes severe neurologic dysfunction. A pseudogene for this gene has been identified on chromosome Y, near the sex determining region Y (SRY) gene. [provided by RefSeq]
Sequence: MESGDEAAIERHRVHLRSATLRDAVPATLHLLPCEVAVDGPAPVGRFFTPAIRQGPEGLEVSFRGRCLRGEEVAVPPGLVGYVMVTEEKKVSMGKPDPLRDSGTDDQEEEPLERDFDRFIGATANFSRFTLWGLETIPGPDAKVRGALTWPSLAAAIHAQVPEDSpecifications
NP_115569.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MESGDEAAIERHRVHLRSATLRDAVPATLHLLPCEVAVDGPAPVGRFFTPAIRQGPEGLEVSFRGRCLRGEEVAVPPGLVGYVMVTEEKKVSMGKPDPLRDSGTDDQEEEPLERDFDRFIGATANFSRFTLWGLETIPGPDAKVRGALTWPSLAAAIHAQVPED | |
RUO | |
RNASEH2C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
84153 | |
AYP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AGS3/AYP1/FLJ20974/MGC22934 | |
RNASEH2C | |
Recombinant | |
wheat germ expression system |