missing translation for 'onlineSavingsMsg'
Läs mer

R&D Systems™ Human B3GalT4 (NP_003773) VersaClone cDNA

Produktkod. 18700319
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18700319 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18700319 Leverantör R&D Systems™ Leverantörsnummer RDC2263

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

R&D Systems™ Human B3GalT4 (NP_003773) VersaClone cDNA is a member of the beta-1,3-galactosyltransferase (beta3GalT) gene family.

The Human B3GalT4 (NP_003773) VersaClone cDNA clone contains the complete ORF for B3GALT4, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The B3GALT4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY

Specifikationer

Gen B3GALT4
Tillträdesnummer NP_003773
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10 mM Tris-Cl
Infoga typ 1150bp
pH 8.5
Gen-ID (Entrez) 8705
Gene Alias b3Gal-T4, beta-1,3-galactosyltransferase 4, Beta-1,3-GalTase 4, beta-3-galactosyltransferase 4, Beta3GalT4, Beta3Gal-T4, EC 2.4.1.62, Gal-T2, GALT2, GalT4, Ganglioside galactosyltransferase, UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 4, UDP-galactose:beta-N-acetyl-galactosamine-beta-1,3-galactosyltransferase
Innehåll och lagring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Art Human
Kvantitet 10 μg
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.