Learn More
Abnova™ Human B3GALTL Full-length ORF (NP_919299.2, 1 a.a. - 498 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00145173-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a beta-1,3-glucosyltransferase that transfers glucose to O-linked fucosylglycans on thrombospondin type-1 repeats (TSRs) of several proteins. The encoded protein is a type II membrane protein. Defects in this gene are a cause of Peters-plus syndrome (PPS)
Sequence: MRPPACWWLLAPPALLALLTCSLAFGLASEDTKKEVKQSQDLEKSGISRKNDIDLKGIVFVIQSQSNSFHAKRAEQLKKSILKQAADLTQELPSVLLLHQLAKQEGAWTILPLLPHFSVTYSRNSSWIFFCEEETRIQIPKLLETLRRYDPSKEWFLGKALHDEEATIIHHYAFSENPTVFKYPDFAAGWALSIPLVNKLTKRLKSESLKSDFTIDLKHEIALYIWDKGGGPPLTPVPEFCTNDVDFYCATTFHSFLPLCRKPVKKKDIFVAVKTCKKFHGDRIPIVKQTWESQASLIEYYSDYTENSIPTVDLGIPNTDRGHCGKTFAILERFLNRSQDKTAWLVIVDDDTLISISRLQHLLSCYDSGKPVFLGERYGYGLGTGGYSYITGGGGMVFSREAVRRLLASKCRCYSNDAPDDMVLGMCFSGLGIPVTHSPLFHQARPVDYPKDYLSHQVPISFHKHWNIDPVKVYFTWLAPSDEDKARQETQKGFREELSpecifications
NP_919299.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
83kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
B3GALTL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
145173 | |
B3GALTL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
B3GTL/B3Glc-T/Gal-T/beta3Glc-T | |
B3GALTL | |
Recombinant | |
wheat germ expression system |