Learn More
Abnova™ Human B4GAT1 Partial ORF (NP_006867.1, 316 a.a. - 415 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011041-Q01.25ug
Dettagli aggiuntivi : Weight : 0.00010kg
Descrizione
This gene encodes a member of the beta-1,3-N-acetylglucosaminyltransferase family. This enzyme is a type II transmembrane protein. It is essential for the synthesis of poly-N-acetyllactosamine, a determinant for the blood group i antigen. [provided by RefSeq]
Sequence: AYVVPWQDPWEPFYVAGGKVPTFDERFRQYGFNRISQACELHVAGFDFEVLNEGFLVHKGFKEALKFHPQKEAENQHNKILYRQFKQELKAKYPNSPRRCSpecifica
NP_006867.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AYVVPWQDPWEPFYVAGGKVPTFDERFRQYGFNRISQACELHVAGFDFEVLNEGFLVHKGFKEALKFHPQKEAENQHNKILYRQFKQELKAKYPNSPRRC | |
RUO | |
B4GAT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11041 | |
B4GAT1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
B3GN-T1/B3GNT6/BETA3GNTI/iGAT/iGNT | |
B4GAT1 | |
Recombinant | |
wheat germ expression system |