missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BAIAP2L1 Partial ORF (NP_061330, 398 a.a. - 511 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055971-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16178556

  • 3343.96 SEK / 10µg

Estimated Shipment: 09-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: KLLEENETEAVTVPTPSPTPVRSISTVNLSENSSVVIPPPDYLECLSMGAAADRRADSARTTSTFKAPASKPETAAPNDANGTAKPPFLSGENPFATVKLRPTVTNDRSAPIIR
Specifications

Specifications

NP_061330
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.28kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KLLEENETEAVTVPTPSPTPVRSISTVNLSENSSVVIPPPDYLECLSMGAAADRRADSARTTSTFKAPASKPETAAPNDANGTAKPPFLSGENPFATVKLRPTVTNDRSAPIIR
RUO
BAIAP2L1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
55971
BAIAP2L1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IRTKS
BAIAP2L1
Yes
Videos
SDS
Documents
Certificates