missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BAT3 Full-length ORF (NP_542433.1, 1 a.a. - 1126 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_542433.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7917 |
Molecular Weight (g/mol) | 145.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16114538
|
Abnova™
H00007917-P02.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16104538
|
Abnova™
H00007917-P02.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MEPNDSTSTAVEEPDSLEVLVKTLDSQTRTFIVGAQMNVKEFKEHIAASVSIPSEKQRLIYQGRVLQDDKKLQEYNVGGKVIHLVERAPPQTHLPSGASSGTGSASATHGGGSPPGTRGPGASVHDRNANSYVMVGTFNLPSDGSAVDVHINMEQAPIQSEPRVRLVMAQHMIRDIQTLLSRMECRGGPQPQHSQPPPQPPAVTPEPVALSSQTSEPVESEAPPREPMEAEEVEERAPAQNPELTPGPAPAGPTPAPETNAPNHPSPAEYVEVLQELQRLESRLQPFLQRYYEVLGAAATTDYNNNHEGREEDQRLINLVGESLRLLGNTFVALSDLRCNLACTPPRHLHVVRPMSHYTTPMVLQQAAIPIQINVGTTVTMTGNGTRPPPTPNAEAPPPGPGQASSVAPSSTNVESSAEGAPPPGPAPPPATSHPRVIRISHQSVEPVVMMHMNIQDSGTQPGGVPSAPTGPLGPPGHGQTLGQQVPGFPTAPTRVVIARPTPPQARPSHPGGPPVSGTLQGAGLGTNASLAQMVSGLVGQLLMQPVLVAQGTPGMAPPPAPATASASAGTTNTATTAGPAPGGPAQPPPTPQPSMADLQFSQLLGNLLGPAGPGAGGSGVASPTITVAMPGVPAFLQGMTDFLQATQTAPPPPPPPPPPPPAPEQQTMPPPGSPSGGAGSPGGLGLESLSPEFFTSVVQGVLSSLLGSLGARAGSSESIAAFIQRLSGSSNIFEPGADGALGFFGALLSLLCQNFSMVDVVMLLHGHFQPLQRLQPQLRSFFHQHYLGGQEPTPSNIRMATHTLITGLEEYVRESFSLVQVQPGVDIIRTNLEFLQEQFNSIAAHVLHCTDSGFGARLLELCNQGLFECLALNLHCLGGQQMELAAVINGRIRRMSRGVNPSLVSWLTTMMGLRLQVVLEHMPVGPDAILRYVRRVGDPPQPLPEEPMEVQGAERASPEPQRENASPAPGTTAEEAMSRGPPPAPEGGSRDEQDGASAETEPWAAAVPPEWVPIIQQDIQSQRKVKPQPPLSDAYLSGMPAKRRKTMQGEGPQLLLSEAVSRAAKAAGARPLTSPESLSRDLEAPEVQESYRQQLRSDIQKRLQEDPNYSPQRFPNAQRAFADDPSpecifications
NP_542433.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
145.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BAG-6/BAG6/D6S52E/G3 | |
BAT3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7917 | |
BAT3 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
BAT3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |