missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BAT5 Partial ORF (NP_066983, 459 a.a. - 556 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4055.00 SEK - 6150.00 SEK
Specifications
Name | BAT5 (Human) Recombinant Protein (Q01) |
---|---|
Recombinant | Yes |
For Use With (Application) | Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) |
Molecular Weight | 36.52kDa |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16128075
|
Abnova™
H00007920-Q01.25UG |
25 ug |
6150.00 SEK
25µg |
Estimated Shipment: 11-08-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
16118075
|
Abnova™
H00007920-Q01.10UG |
10 ug |
4055.00 SEK
10µg |
Estimated Shipment: 11-08-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Description
A cluster of genes, BAT1-BAT5, has been localized in the vicinity of the genes for TNF alpha and TNF beta. These genes are all within the human major histocompatibility complex class III region. The protein encoded by this gene is thought to be involved in some aspects of immunity. [provided by RefSeq]
Sequence: PRVMAEEGLRVVRQWLEASSQLEEASIYSRWEVEEDWCLSVLRSYQAEHGPDFPWSVGEDMSADGRRQLALFLARKHLHNFEATHCTPLPAQNFQMPWSpecifications
BAT5 (Human) Recombinant Protein (Q01) | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
36.52kDa |