missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BBOX1 Full-length ORF (AAH11034, 1 a.a. - 387 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008424-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16132442

  • 6845.00 SEK / 25µg
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes gamma butyrobetaine hydroxylase which catalyzes the formation of L-carnitine from gamma-butyrobetaine, the last step in the L-carnitine biosynthetic pathway. Carnitine is essential for the transport of activated fatty acids across the mitochondrial membrane during mitochondrial beta-oxidation. [provided by RefSeq]

Sequence: MACTIQKAEALDGAHLMQILWYDEEESLYPAVWLRDNCPCSDCYLDSAKARKLLVEALDVNIGIKGLIFDRKKVYITWPDEHYSEFQADWLKKRCFSKQARAKLQRELFFPECQYWGSELQLPTLDFEDVLRYDEHAYKWLSTLKKVGIVRLTGASDKPGEVSKLGKRMGFLYLTFYGHTWQVQDKIDANNVAYTTGKLSFHTDYPALHHPPGVQLLHCIKQTVTGGDSEIVDGFNVCQKLKKNNPQAFQILSSTFVDFTDIGVDYCDFSVQSKHKIIELDDKGQVVRINFNNATRDTIFDVPVERVQPFYAALKEFVDLMNSKESKFTFKMNPGDVITFDNWRLLHGRRSYEAGTEISRHLEGAYADWDVVMSRLRILRQRVENGN
Specifications

Specifications

AAH11034
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
68.31kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
BBOX1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
8424
BBOX1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BBH/BBOX/G-BBH/gamma-BBH
BBOX1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers