missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BBS1 Partial ORF (NP_078925, 187 a.a. - 295 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000582-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195154

  • 6845.00 SEK / 25µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Mutations in this gene have been observed in patients with the major form (type 1) of Bardet-Biedl syndrome. The encoded protein may play a role in eye, limb, cardiac and reproductive system development. [provided by RefSeq]

Sequence: NQHKSNSIKRQTVITTMTTLKKNLADEDAVSCLVLGTENKELLVLDPEAFTILAKMSLPSVPVFLEVSGQFDVEFRLAAACRNGNIYILRRDSKHPKYCIELSAQPVGL
Specifications

Specifications

NP_078925
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NQHKSNSIKRQTVITTMTTLKKNLADEDAVSCLVLGTENKELLVLDPEAFTILAKMSLPSVPVFLEVSGQFDVEFRLAAACRNGNIYILRRDSKHPKYCIELSAQPVGL
RUO
BBS1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
582
BBS1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BBS2L2/FLJ23590/MGC126183/MGC126184/MGC51114
BBS1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers