Learn More
Abnova™ Human BCAS1 Full-length ORF (ADR83077.1, 1 a.a. - 584 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | ADR83077.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8537 |
Molecular Weight (g/mol) | 90.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16162542
|
Abnova™
H00008537-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16152542
|
Abnova™
H00008537-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene resides in a region at 20q13 which is amplified in a variety of tumor types and associated with more aggressive tumor phenotypes. Among the genes identified from this region, it was found to be highly expressed in three amplified breast cancer cell lines and in one breast tumor without amplification at 20q13.2. However, this gene is not in the common region of maximal amplification and its expression was not detected in the breast cancer cell line MCF7, in which this region is highly amplified. Although not consistently expressed, this gene is a candidate oncogene. [provided by RefSeq]
Sequence: MGNQMSVPQRVEDQENEPEAETYKDNASALNGVPVVVSTHTVQHLEEVDLGISVKTDNVATSSPETTEISAVADANGKNLGKEAKPEAPAAKSRFFLMLSRPVPGRTGDQAADSSLGSVKLDVSSNKAPANKDPSESWTLPVAAGPGQDTDKTPGHAPAQDKALSAARDPTLLPPETGGAGGEAPSKPKDSSFFDKFFKLDKGQEKVPGDSQQEAKRAEHQDKVDEVPGLSGQSDDVPAGKDIVDGKEKEGQELGTADCSVPGDPEGLETAKDDSQAAAIAENNNSIMSFFKTLVSPNKAETKKDPEDTGAEKSPTTSADLKSDKANFTSQETQGAGKNSKGCNPSGHTQSVTTPEPAKEGTKEKSGPTSLPLGKLFWKKSVKEDSVPTGAEENVVCESPVEIIKSKEVESALQTVDLNEGDAAPEPTEAKLKREESKPRTSLMAFLRQMSVKGDGGITHSEEINGKDSSCQTSDSTEKTITPPEPEPTGAPQKGKEGSSKDKKSAAEMNKQKSNKQEAKEPAQCTEQATVDTNSLQNGDKLQKRPEKRQQSLGGFFKGLGPKRMLDAQVQTDPVSIGPVGKSKSpecifications
ADR83077.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
90.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AIBC1/NABC1 | |
BCAS1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8537 | |
BCAS1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
RUO | |
BCAS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |