missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BCL3 Full-length ORF (NP_005169.1, 1 a.a. - 446 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000602-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16132181

  • 4515.00 SEK / 10µg
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a proto-oncogene candidate. It is identified by its translocation into the immunoglobulin alpha-locus in some cases of B-cell leukemia. The protein encoded by this gene contains seven ankyrin repeats, which are most closely related to those found in I kappa B proteins. This protein functions as a transcriptional co-activator that activates through its association with NF-kappa B homodimers. The expression of this gene can be induced by NF-kappa B, which forms a part of the autoregulatory loop that controls the nuclear residence of p50 NF-kappa B. [provided by RefSeq]

Sequence: MDEGPVDLRTRPKAAGLPGAALPLRKRPLRAPSPEPAAPRGAAGLVVPLDPLRGGCDLPAVPGPPHGLARPEALYYPGALLPLYPTRAMGSPFPLVNLPTPLYPMMCPMEHPLSADIAMATRADEDGDTPLHIAVVQGNLPAVHRLVNLFQQGGRELDIYNNLRQTPLHLAVITTLPSVVRLLVTAGASPMALDRHGQTAAHLACEHRSPTCLRALLDSAAPGTLDLEARNYDGLTALHVAVNTECQETVQLLLERGADIDAVDIKSGRSPLIHAVENNSLSMVQLLLQHGANVNAQMYSGSSALHSASGRGLLPLVRTLVRSGADSSLKNCHNDTPLMVARSRRVIDILRGKATRPASTSQPDPSPDRSANTSPESSSRLSSNGLLSASPSSSPSQSPPRDPPGFPMAPPNFFLPSPSPPAFLPFAGVLRGPGRPVPPSPAPGGS
Specifications

Specifications

NP_005169.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
BCL3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
602
BCL3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BCL4/D19S37
BCL3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers