Learn More
Abnova™ Human BHMT2 Full-length ORF (NP_060084.2, 1 a.a. - 363 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023743-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Homocysteine is a sulfur-containing amino acid that plays a crucial role in methylation reactions. Transfer of the methyl group from betaine to homocysteine creates methionine, which donates the methyl group to methylate DNA, proteins, lipids, and other intracellular metabolites. The protein encoded by this gene is one of two methyl transferases that can catalyze the transfer of the methyl group from betaine to homocysteine. Anomalies in homocysteine metabolism have been implicated in disorders ranging from vascular disease to neural tube birth defects such as spina bifida. [provided by RefSeq]
Sequence: MAPAGRPGAKKGILERLESGEVVIGDGSFLITLEKRGYVKAGLWTPEAVIEHPDAVRQLHMEFLRAGSNVMQTFTFSASEDNMESKWEDVNAAACDLAREVAGKGDALVAGGICQTSIYKYQKDEARIKKLFRQQLEVFAWKNVDFLIAEYFEHVEEAVWAVEVLKESDRPVAVTMCIGPEGDMHDITPGECAVRLVKAGASIVGVNCRFGPDTSLKTMELMKEGLEWAGLKAHLMVQPLGFHAPDCGKEGFVDLPEYPFGLESRVATRWDIQKYAREAYNLGVRYIGGCCGFEPYHIRAIAEELAPERGFLPPASEKHGSWGSGLDMHTKPWIRARARREYWENLLPASGRPFCPSLSKPDFSpecifications
NP_060084.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
BHMT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
23743 | |
BHMT2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20001 | |
BHMT2 | |
Recombinant | |
wheat germ expression system |