missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BIRC6 Partial ORF (NP_057336, 31 a.a. - 140 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_057336 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 57448 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16162683
|
Abnova™
H00057448-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16172683
|
Abnova™
H00057448-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein with a BIR (baculoviral inhibition of apoptosis protein repeat) domain and a UBCc (ubiquitin-conjugating enzyme E2, catalytic) domain. This protein inhibits apoptosis by facilitating the degradation of apoptotic proteins by ubiquitination. [provided by RefSeq]
Sequence: RDGCMHCDADGLHSLSYHPALNAILAVTSRGTIKVIDGTSGATLQASALSAKPGGQVKCQYISAVDKVIFVDDYAVGCRKDLNGILLLDTALQTPVSKQDDVVQLELPVTSpecifications
NP_057336 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APOLLON/BRUCE/FLJ13726/FLJ13786/KIAA1289 | |
BIRC6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
57448 | |
BIRC6 (Human) Recombinant Protein (Q01) | |
RDGCMHCDADGLHSLSYHPALNAILAVTSRGTIKVIDGTSGATLQASALSAKPGGQVKCQYISAVDKVIFVDDYAVGCRKDLNGILLLDTALQTPVSKQDDVVQLELPVT | |
RUO | |
BIRC6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |