Learn More
Abnova™ Human BMP10 Partial ORF (NP_055297, 317 a.a. - 424 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00027302-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the TGF-beta family of growth factors. Data suggest that the similar protein in mouse plays an important role in trabeculation of the embryonic heart. In human, this protein may signal through receptor serine/threonine kinases. [provided by RefSeq]
Sequence: NAKGNYCKRTPLYIDFKEIGWDSWIIAPPGYEAYECRGVCNYPLAEHLTPTKHAIIQALVHLKNSQKASKACCVPTKLEPISILYLDKGVVTYKFKYEGMAVSECGCRSpecifications
NP_055297 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NAKGNYCKRTPLYIDFKEIGWDSWIIAPPGYEAYECRGVCNYPLAEHLTPTKHAIIQALVHLKNSQKASKACCVPTKLEPISILYLDKGVVTYKFKYEGMAVSECGCR | |
RUO | |
BMP10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
27302 | |
BMP10 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC126783 | |
BMP10 | |
Recombinant | |
wheat germ expression system |