missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BMP8A Partial ORF (NP_861525.1, 112 a.a. - 208 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00353500-Q01.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16075825

  • 6845.00 SEK / 25µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: VERDRALGHQEPHWKEFRFDLTQIPAGEAVTAAEFRIYKVPSIHLLNRTLHVSMFQVVQEQSNRESDLFFLDLQTLRAGDEGWLVLDVTAASDCWLL
Specifications

Specifications

NP_861525.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VERDRALGHQEPHWKEFRFDLTQIPAGEAVTAAEFRIYKVPSIHLLNRTLHVSMFQVVQEQSNRESDLFFLDLQTLRAGDEGWLVLDVTAASDCWLL
RUO
BMP8A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
353500
BMP8A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ14351/FLJ45264
BMP8A
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers