missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BMP8A Partial ORF (NP_861525.1, 112 a.a. - 208 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00353500-Q01.10ug
Additional Details : Weight : 0.02000kg
Description
Sequence: VERDRALGHQEPHWKEFRFDLTQIPAGEAVTAAEFRIYKVPSIHLLNRTLHVSMFQVVQEQSNRESDLFFLDLQTLRAGDEGWLVLDVTAASDCWLLSpecifications
NP_861525.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VERDRALGHQEPHWKEFRFDLTQIPAGEAVTAAEFRIYKVPSIHLLNRTLHVSMFQVVQEQSNRESDLFFLDLQTLRAGDEGWLVLDVTAASDCWLL | |
RUO | |
BMP8A | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
353500 | |
BMP8A (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ14351|FLJ45264 | |
BMP8A | |
Yes |