Learn More
Abnova™ Human BPGM Full-length ORF (AAH17050, 1 a.a. - 259 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000669-P01.10ug
Additional Details : Weight : 0.00010kg
Description
2,3-diphosphoglycerate (2,3-DPG) is a small molecule found at high concentrations in red blood cells where it binds to and decreases the oxygen affinity of hemoglobin. This gene encodes a multifunctional enzyme that catalyzes 2,3-DPG synthesis via its synthetase activity, and 2,3-DPG degradation via its phosphatase activity. The enzyme also has phosphoglycerate phosphomutase activity. Deficiency of this enzyme increases the affinity of cells for oxygen. Mutations in this gene result in hemolytic anemia. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]
Sequence: MSKYKLIMLRHGEGAWNKENRFCSWVDQKLNSEGMEEARNCGKQLKALNFEFDLVFTSVLNRSIHTAWLILEELGQEWVPVESSWRLNERHYGALIGLNREQMALNHGEEQVRLWRRSYNVTPPPIEESHPYYQEIYNDRRYKVCDVPLDQLPRSESLKDVLERLLPYWNERIAPEVLRGKTILISAHGNSSRALLKHLEGISDEDIINITLPTGVPILLELDENLRAVGPHQFLGDQEAIQAAIKKVEDQGKVKQAKKSpecifications
AAH17050 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.23kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
BPGM | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
669 | |
BPGM (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BPGM | |
Wheat Germ (in vitro) | |
GST | |
Liquid |