Läs mer
Novus Biologicals™ Human Brain Pons Whole Tissue Lysate (Adult Whole Alzheimer's)
Handla allt Bio Techne ProdukterBeskrivning
The Human Brain Pons Whole Tissue Lysate (Adult Whole Alzheimer's) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifikationer
Specifikationer
| För användning med (applikation) | Immunoprecipitation, Western Blot |
| Vävnad | Nervous |
| Kvantitet | 1 mg |
| Förvaringsbuffert | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Testspecificitet | Total Protein - Alzheimers Disease: Brain: Pons |
| Cellulär fraktion | Whole |
| Vävnadsskick | Alzheimer's |
| Korsreaktivitet | Human |
| Värd art | Human |
| Koncentration | 5 mg/mL |
| Visa mer |
For Research Use Only