Learn More
Abnova™ Human BTBD2 Partial ORF (NP_060267, 422 a.a. - 503 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055643-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The C-terminus of the protein encoded by this gene binds topoisomerase I. The N-terminus contains a proline-rich region and a BTB/POZ domain (broad-complex, Tramtrack and bric a brac/Pox virus and Zinc finger), both of which are typically involved in protein-protein interactions. Subcellularly, the protein localizes to cytoplasmic bodies. [provided by RefSeq]
Sequence: IQIIHTDSNTVLGQNDTGFSCDGSASTFRVMFKEPVEVLPNVNYTACATLKGPDSHYGTKGLRKVTHESPTTGAKTCFTFCYSpecifications
NP_060267 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.76kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IQIIHTDSNTVLGQNDTGFSCDGSASTFRVMFKEPVEVLPNVNYTACATLKGPDSHYGTKGLRKVTHESPTTGAKTCFTFCY | |
RUO | |
BTBD2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
55643 | |
BTBD2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BTBD2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |