missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BTBD9 Full-length ORF (AAI01358.1, 1 a.a. - 582 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00114781-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16127093

  • 6845.00 SEK / 25µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MCRALLYGGMRESQPEAEIPLQDTTAEAFTMLLKYIYTGRATLTDEKEEVLLDFLSLAHKYGFPELEDSTSEYLCTILNIQNVCMTFDVASLYSLPKLTCMCCMFMDRNAQEVLSSEGFLSLSKTALLNIVLRDSFAAPEKDIFLALLNWCKHNSKENHAEIMQAVRLPLMSLTELLNVVRPSGLLSPDAILDAIKVRSESRDMDLNYRGMLIPEENIATMKYGAQVVKGELKSALLDGDTQNYDLDHGFSRHPIDDDCRSGIEIKLGQPSIINHIRILLWDRDSRSYSYFIEVSMDELDWVRVIDHSQYLCRSWQKLYFPARVCSGDGVSLWCPLWSRTPELKQSSLLGLPKCRYIRIVGTHNTVNKIFHIVAFECMFTNKTFTLEKGLIVPMENVATIADCASVIEGVSRSRNALLNGDTKNYDWDSGYTCHQLGSGAIVVQLAQPYMIGSIRLLLWDCDDRSYSYYVEVSTNQQQWTMVADRTKVSCKSWQSVTFERQPASFIRIVGTHNTANEVFHCVHFECPEQQSSQKEENSEESGTGDTSLAGQQLDSHALRAPSGSSLPSSPGSNSRSPNRQHQ
Specifications

Specifications

AAI01358.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
92.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
BTBD9
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
114781
BTBD9 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ32945/KIAA1880/MGC120517/MGC120519/MGC120520/dJ322I12.1
BTBD9
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers