missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BTN3A2 Full-length ORF (NP_008978.2, 1 a.a. - 334 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_008978.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 11118 |
Molecular Weight (g/mol) | 62.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16136192
|
Abnova™
H00011118-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16146192
|
Abnova™
H00011118-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the immunoglobulin superfamily, containing two Ig domains with similarity to Ig variable and Ig constant domains. The gene resides in the juxta-telomeric region of the major histocompatability class 1 locus on chromosome 6 in the seven member BTN cluster, which includes butyrophilin, and three members each of the BTN2 and BTN3 subfamilies. Alternatively spliced transcript variants have been described but their full-length nature has yet to be determined. [provided by RefSeq]
Sequence: MKMASSLAFLLLNFHVSLLLVQLLTPCSAQFSVLGPSGPILAMVGEDADLPCHLFPTMSAETMELKWVSSSLRQVVNVYADGKEVEDRQSAPYRGRTSILRDGITAGKAALRIHNVTASDSGKYLCYFQDGDFYEKALVELKVAALGSNLHVEVKGYEDGGIHLECRSTGWYPQPQIQWSNAKGENIPAVEAPVVADGVGLYEVAASVIMRGGSGEGVSCIIRNSLLGLEKTASISIADPFFRSAQPWIAALAGTLPILLLLLAGASYFLWRQQKEITALSSEIESEQEMKEMGYAATEREISLRESLQEELKRKKIQYLTRGEESSSDTNKSASpecifications
NP_008978.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BT3.2/BT3.3/BTF4 | |
BTN3A2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
11118 | |
BTN3A2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
BTN3A2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |