missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C13orf7 Partial ORF (NP_078822, 1 a.a. - 95 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079596-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16134243

  • 3343.96 SEK / 10µg

Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MLSHTVRKHLRKTRLELLHKEYEDEIDCLQKEVEELKSKNLSLESQIKTILDPLTLVQGNQNEDKHLVTDNPSKINPETVAEWKKKLRTANEIYE
Specifications

Specifications

NP_078822
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.19kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MLSHTVRKHLRKTRLELLHKEYEDEIDCLQKEVEELKSKNLSLESQIKTILDPLTLVQGNQNEDKHLVTDNPSKINPETVAEWKKKLRTANEIYE
RUO
RNF219
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
79596
C13orf7 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C13orf7/DKFZp686A01276/DKFZp686N15250/DKFZp686O03173/FLJ13449/FLJ25774
RNF219
Yes
Videos
SDS
Documents
Certificates