Learn More
Abnova™ Human C15orf16 Partial ORF (NP_570971, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_570971 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 161725 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16172247
|
Abnova™
H00161725-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16162247
|
Abnova™
H00161725-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Deubiquitinating enzymes (DUBs; see MIM 603478) are proteases that specifically cleave ubiquitin (MIM 191339) linkages, negating the action of ubiquitin ligases. OTUD7A belongs to a DUB subfamily characterized by an ovarian tumor (OTU) domain.[supplied by OMIM]
Sequence: MVSSVLPNPTSAECWAALLHDPMTLDMDAVLSDFVRSTGAEPGLARDLLEGKNWDLTAALSDYEQLRQVHTANLPHVFNEGRGPKQPEREPQPGHKVERSpecifications
NP_570971 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C15orf16/C16ORF15/CEZANNE2/OTUD7 | |
OTUD7A | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
161725 | |
C15orf16 (Human) Recombinant Protein (Q01) | |
MVSSVLPNPTSAECWAALLHDPMTLDMDAVLSDFVRSTGAEPGLARDLLEGKNWDLTAALSDYEQLRQVHTANLPHVFNEGRGPKQPEREPQPGHKVER | |
RUO | |
OTUD7A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |