Learn More
Abnova™ Human C19orf2 Full-length ORF (NP_604431.1, 1 a.a. - 495 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_604431.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8725 |
Molecular Weight (g/mol) | 82kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16192812
|
Abnova™
H00008725-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16182812
|
Abnova™
H00008725-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene binds to RNA polymerase II subunit 5 (RPB5) and negatively modulates transcription through its binding to RPB5. The encoded protein seems to have inhibitory effects on various types of activated transcription, but it requires the RPB5-binding region. This protein acts as a corepressor. It is suggested that it may require signaling processes for its function or that it negatively modulates genes in the chromatin structure. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]
Sequence: MWRGCARSRKRKKVDNDYNALRERLSTLPDKLSYNIMVPFGPFAFMPGKLVHTNEVTVLLGDNWFAKCSAKQAVGLVEHRKEHVRKTIDDLKKVMKNFESRVEFTEDLQKMSDAAGDIVDIREEIKCDFEFKAKHRIAHKPHSKPKTSDIFEADIANDVKSKDLLADKELWARLEELERQEELLGELDSKPDTVIANGEDTTSSEEEKEDRNTNVNAMHQVTDSHTPCHKDVASSEPFSGQVNSQLNCSVNGSSSYHSDDDDDDDDDDDDDNIDDDDGDNDHEALGVGDNSIPTIYFSHTVEPKRVRINTGKNTTLKFSEKKEEAKRKRKNSTGSGHSAQELPTIRTPADIYRAFVDVVNGEYVPRKSILKSRSRENSVCSDTSESSAAEFDDRRGVLRSISCEEATCSDTSESILEEEPQENQKKLLPLSVTPEAFSGTVIEKEFVSPSLTPPPAIAHPALPTIPERKEVLLEASEETGKRVSKFKAARLQQKDSpecifications
NP_604431.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
82kDa | |
Glutathione Sepharose 4 Fast Flow | |
MWRGCARSRKRKKVDNDYNALRERLSTLPDKLSYNIMVPFGPFAFMPGKLVHTNEVTVLLGDNWFAKCSAKQAVGLVEHRKEHVRKTIDDLKKVMKNFESRVEFTEDLQKMSDAAGDIVDIREEIKCDFEFKAKHRIAHKPHSKPKTSDIFEADIANDVKSKDLLADKELWARLEELERQEELLGELDSKPDTVIANGEDTTSSEEEKEDRNTNVNAMHQVTDSHTPCHKDVASSEPFSGQVNSQLNCSVNGSSSYHSDDDDDDDDDDDDDNIDDDDGDNDHEALGVGDNSIPTIYFSHTVEPKRVRINTGKNTTLKFSEKKEEAKRKRKNSTGSGHSAQELPTIRTPADIYRAFVDVVNGEYVPRKSILKSRSRENSVCSDTSESSAAEFDDRRGVLRSISCEEATCSDTSESILEEEPQENQKKLLPLSVTPEAFSGTVIEKEFVSPSLTPPPAIAHPALPTIPERKEVLLEASEETGKRVSKFKAARLQQKD | |
RUO | |
C19orf2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
8725 | |
C19orf2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10575/NNX3/RMP/URI | |
C19orf2 | |
Recombinant | |
wheat germ expression system |