missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C19orf2 Full-length ORF (NP_604431.1, 1 a.a. - 495 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008725-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16182812

  • 3343.96 SEK / 10µg

Estimated Shipment: 19-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene binds to RNA polymerase II subunit 5 (RPB5) and negatively modulates transcription through its binding to RPB5. The encoded protein seems to have inhibitory effects on various types of activated transcription, but it requires the RPB5-binding region. This protein acts as a corepressor. It is suggested that it may require signaling processes for its function or that it negatively modulates genes in the chromatin structure. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]

Sequence: MWRGCARSRKRKKVDNDYNALRERLSTLPDKLSYNIMVPFGPFAFMPGKLVHTNEVTVLLGDNWFAKCSAKQAVGLVEHRKEHVRKTIDDLKKVMKNFESRVEFTEDLQKMSDAAGDIVDIREEIKCDFEFKAKHRIAHKPHSKPKTSDIFEADIANDVKSKDLLADKELWARLEELERQEELLGELDSKPDTVIANGEDTTSSEEEKEDRNTNVNAMHQVTDSHTPCHKDVASSEPFSGQVNSQLNCSVNGSSSYHSDDDDDDDDDDDDDNIDDDDGDNDHEALGVGDNSIPTIYFSHTVEPKRVRINTGKNTTLKFSEKKEEAKRKRKNSTGSGHSAQELPTIRTPADIYRAFVDVVNGEYVPRKSILKSRSRENSVCSDTSESSAAEFDDRRGVLRSISCEEATCSDTSESILEEEPQENQKKLLPLSVTPEAFSGTVIEKEFVSPSLTPPPAIAHPALPTIPERKEVLLEASEETGKRVSKFKAARLQQKD
Specifications

Specifications

NP_604431.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
82kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10575/NNX3/RMP/URI
C19orf2
Yes
wheat germ expression system
Liquid
8725
C19orf2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MWRGCARSRKRKKVDNDYNALRERLSTLPDKLSYNIMVPFGPFAFMPGKLVHTNEVTVLLGDNWFAKCSAKQAVGLVEHRKEHVRKTIDDLKKVMKNFESRVEFTEDLQKMSDAAGDIVDIREEIKCDFEFKAKHRIAHKPHSKPKTSDIFEADIANDVKSKDLLADKELWARLEELERQEELLGELDSKPDTVIANGEDTTSSEEEKEDRNTNVNAMHQVTDSHTPCHKDVASSEPFSGQVNSQLNCSVNGSSSYHSDDDDDDDDDDDDDNIDDDDGDNDHEALGVGDNSIPTIYFSHTVEPKRVRINTGKNTTLKFSEKKEEAKRKRKNSTGSGHSAQELPTIRTPADIYRAFVDVVNGEYVPRKSILKSRSRENSVCSDTSESSAAEFDDRRGVLRSISCEEATCSDTSESILEEEPQENQKKLLPLSVTPEAFSGTVIEKEFVSPSLTPPPAIAHPALPTIPERKEVLLEASEETGKRVSKFKAARLQQKD
RUO
C19orf2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates